Assay Details
Gene Symbol: LOC574538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033859.1 | 5 - 6 | 2152 | 94 | ||
GenBank mRNA | AK096077.1 | - | 5 - 6 | 2152 | 94 | |
BC036774.1 | - | 4 - 5 | 4318 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | family with sequence similarity 138 member D | FAM138D | F379 | -102.5kb | Sense | Chr. 12 - 36661 - 38133 on NCBI Build GRCh38 |
Downstream | solute carrier family 6 member 12 | SLC6A12 |
BGT-1, BGT1, GAT2 |
48.8kb | Sense | Chr. 12 - 190077 - 214523 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001076.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12014209.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12014209.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12014209.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12014209.hg.1 | TC12001076.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009542.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200171280.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171280.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171280.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171280.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200171280.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171280.hg.1 | ENST00000505893.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171280.hg.1 | ENST00000508953.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200171280.hg.1 | NR_033859.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200171280.hg.1 | TC1200009542.hg.1 | Clariom D TC |
