Assay Details
| Gene Symbol: NAV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ488204.1 | - | 1 | 251 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4486 | MIR4486 | -21.4kb | Sense | Chr. 11 - 19575310 - 19575372 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100126784 | LOC100126784 | 113.8kb | Anti-sense | Chr. 11 - 19710934 - 19714672 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AJ488204.1 transcripts.
This assay is in the 5'UTR region of the AJ488204.1 transcripts.
Set Membership: |
|

NAV2
Genomic Map