Assay Details
| Gene Symbol: NAV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006718366.3 | XP_006718429.1 | 1 | 83 | 62 | |
| GenBank mRNA | AJ488206.1 | - | 1 | 35 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4694 | MIR4694 | -17.4kb | Anti-sense | Chr. 11 - 19760004 - 19760083 on NCBI Build GRCh38 | |
| Downstream | NAV2 antisense RNA 2 | NAV2-AS2 | 266.3kb | Anti-sense | Chr. 11 - 20043775 - 20049303 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ488206.1, XM_006718366.3 transcripts.
Set Membership: |
|

NAV2
Genomic Map