Assay Details
| Gene Symbol: LOXL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC015656.2 | - | 1 - 2 | 182 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | R3H domain and coiled-coil containing 1 like | R3HCC1L |
C10orf28, GIDRP86, GIDRP88, PSORT |
-7.1kb | Sense | Chr. 10 - 98134608 - 98244897 on NCBI Build GRCh38 |
| Downstream | pyridine nucleotide-disulphide oxidoreductase domain 2 | PYROXD2 |
C10orf33, FP3420 |
127.0kb | Anti-sense | Chr. 10 - 98383565 - 98415221 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOXL4
Genomic Map