Assay Details
| Gene Symbol: TRHDE-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026836.1 | 1 - 2 | 748 | 85 | ||
| GenBank mRNA | BC040230.1 | - | 1 - 2 | 723 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tryptophan hydroxylase 2 | TPH2 |
ADHD7, NTPH |
-236.5kb | Anti-sense | Chr. 12 - 71938846 - 72032441 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101928137 | LOC101928137 | 885.7kb | Anti-sense | Chr. 12 - 73159190 - 73208317 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001731.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000261180.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000547300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000547300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000547300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000547300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000547300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549138.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000549922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000552503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000552503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000552503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000552503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | ENST00000552503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012668.hg.1 | NM_013381.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000426250.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000435350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000549957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000550334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000550334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000550334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000550334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | ENST00000550334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011269.hg.1 | NR_026837.1 | Refseq |

TRHDE-AS1
Genomic Map