Assay Details
Gene Symbol: LOC340113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033832.1 | 5 - 6 | 910 | 86 | ||
GenBank mRNA | BC044935.1 | - | 5 - 6 | 910 | 86 | |
BC143768.1 | - | 4 - 5 | 450 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | natriuretic peptide receptor 3 | NPR3 |
ANP-C, ANPR-C, ANPRC, C5orf23, GUCY2B, NPR-C, NPRC |
-169.4kb | Sense | Chr. 5 - 32689076 - 32791724 on NCBI Build GRCh38 |
Downstream | threonyl-tRNA synthetase | TARS | ThrRS | 478.2kb | Sense | Chr. 5 - 33440696 - 33468091 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000145.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029643 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859214 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029691 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029701 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029703 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859230 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029677 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029737 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029714 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029640 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859169 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029742 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029753 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859170 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029692 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859246 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859144 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029642 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859220 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029682 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029667 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029729 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029664 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859193 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029688 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029702 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15680191 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029735 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859174 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029747 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859234 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029660 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859189 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859211 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859121 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859212 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029733 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029661 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029696 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859216 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029680 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029644 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029740 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859206 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029649 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029734 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859145 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859185 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029705 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859225 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029645 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859118 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029683 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859183 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029745 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029707 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859217 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859240 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029662 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029739 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029665 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029746 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859232 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029693 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029743 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029672 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029655 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029681 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859184 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029713 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029670 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859155 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029708 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859208 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029641 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029748 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859142 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859204 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029727 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029730 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029700 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029648 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029671 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859213 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859180 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029751 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859139 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859248 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859199 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029749 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029731 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24029650 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03859181 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3285779 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 24613 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P17080990 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P15152130 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P17080990 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3285779 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3285779 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05001472.hg.1 | TC05000145.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05001472.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05001472.hg.1 | ENST00000507693.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05001472.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000507693.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000507693.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000507693.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000507693.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | ENST00000507693.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05001473.hg.1 | TC05000145.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05001473.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05001473.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500147804.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500147804.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500147804.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500147804.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500147804.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500147804.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500147804.hg.1 | TC0500007081.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000507693.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000507693.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000507693.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000507693.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | ENST00000507693.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007081.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500147803.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500147803.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500147803.hg.1 | ENST00000507693.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500147803.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500147803.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500147803.hg.1 | ENST00000505518.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500147803.hg.1 | ENST00000507693.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500147803.hg.1 | NR_033832.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500147803.hg.1 | TC0500007081.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A028081 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A028081 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1794866 | NR_033832.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1794866 | BC044935.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1794866 | NR_033832.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1794866 | BC044935.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1794866 | NR_033832.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1794866 | BC044935.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1794866 | NR_033832.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1794866 | BC044935.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1794866 | NR_033832.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1794866 | BC044935.1 | human_transcriptome |
