Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
240518_PM_at |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
1570572_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010231.hg.1 |
ENST00000473753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010231.hg.1 |
ENST00000473753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010231.hg.1 |
ENST00000473753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010231.hg.1 |
ENST00000473753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010231.hg.1 |
ENST00000617468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010231.hg.1 |
ENST00000617468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010231.hg.1 |
ENST00000617468.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010231.hg.1 |
ENST00000473753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010231.hg.1 |
ENST00000473753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010231.hg.1 |
ENST00000473753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010231.hg.1 |
ENST00000473753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010231.hg.1 |
ENST00000617468.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010231.hg.1 |
ENST00000617468.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010231.hg.1 |
ENST00000617468.1 |
Ensembl |
Human Genome U133 Plus 2.0 Array |
1570572_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
240518_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
240518_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1570572_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1570572_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
240518_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
240518_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1570572_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
240518_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1570572_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1570572_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
240518_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
240518_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1570572_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1570572_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
240518_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
240518_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1570572_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
240518_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
240518_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1570572_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
240518_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
240518_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1570572_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1570572_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1570572_at9 |
- |
- |
Human Genome U95 Set |
89662_PM_at |
- |
- |
Human Genome U95 Set |
89662_at |
- |
- |
Human Genome U133 Set |
240518_PM_at |
- |
- |
Human Genome U133 Set |
240518_at |
- |
- |