Assay Details
| Gene Symbol: LOC100131821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123052.1 | - | 1 | 2341 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RARA antisense RNA 1 | RARA-AS1 | -4.9kb | Sense | Chr. 17 - 40340867 - 40343136 on NCBI Build GRCh38 | |
| Downstream | gap junction protein delta 3 | GJD3 |
CX31.9, Cx30.2, GJA11, GJC1 |
10.1kb | Sense | Chr. 17 - 40360652 - 40364693 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100131821
Genomic Map