Assay Details
| Gene Symbol: CDK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123134.1 | - | 1 | 1649 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 133 member B | FAM133B | -56.0kb | Sense | Chr. 7 - 92560758 - 92590394 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101927497 | LOC101927497 | 187.8kb | Anti-sense | Chr. 7 - 92836483 - 92917187 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CDK6
Genomic Map