Assay Details
| Gene Symbol: LRRC8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123611.1 | - | 1 | 3152 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kynurenine aminotransferase 1 | KYAT1 |
CCBL1, GTK, KAT1, KATI |
-23.8kb | Sense | Chr. 9 - 128832942 - 128882551 on NCBI Build GRCh38 |
| Downstream | phytanoyl-CoA dioxygenase domain containing 1 | PHYHD1 | 11.1kb | Anti-sense | Chr. 9 - 128920895 - 128942041 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LRRC8A
Genomic Map