Assay Details
| Gene Symbol: LOC100130744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK129556.1 | - | 1 | 1147 | 114 | |
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011514151.2 | XP_011512453.1 | 8 | 8242 | 114 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ANKH inorganic pyrophosphate transport regulator | ANKH |
ANK, CCAL2, CMDJ, CPPDD, HANK, MANK |
Anti-sense | Chr. 5 - 14704800 - 14871778 on NCBI Build GRCh38 | |
| Downstream | microRNA 4637 | MIR4637 | 109.4kb | Anti-sense | Chr. 5 - 14825929 - 14826012 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100130744
Genomic Map