Assay Details
| Gene Symbol: FLJ26086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK129597.1 | - | 1 | 157 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATP synthase, H+ transporting, mitochondrial F1 complex, epsilon subunit pseudogene 2 |
ATP5EP2 | -14.0kb | Anti-sense | Chr. 13 - 27945206 - 27945573 on NCBI Build GRCh38 | |
| Downstream | ureidoimidazoline (2-oxo-4-hydroxy-4-carboxy-5-) decarboxylase | URAD | PRHOXNB | 17.3kb | Sense | Chr. 13 - 27978106 - 27988637 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

FLJ26086
Genomic Map