Assay Details
| Gene Symbol: LOC100129233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130248.1 | - | 1 | 455 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102467213 | LOC102467213 | -410.4kb | Sense | Chr. 5 - 106815197 - 107011014 on NCBI Build GRCh38 | |
| Downstream | F-box and leucine rich repeat protein 17 | FBXL17 |
FBXO13, Fbl17, Fbx13 |
435.8kb | Sense | Chr. 5 - 107859033 - 108382098 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100129233
Genomic Map