Assay Details
| Gene Symbol: RPL9P11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130277.1 | - | 1 | 749 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100506022 | LOC100506022 | -146.8kb | Anti-sense | Chr. 1 - 9425094 - 9428996 on NCBI Build GRCh38 | |
| Downstream | transmembrane protein 201 | TMEM201 | NET5 | 11.4kb | Sense | Chr. 1 - 9588843 - 9614877 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RPL9P11
Genomic Map