Assay Details
| Gene Symbol: ATP9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130839.1 | - | 1 | 152 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100996274 | LOC100996274 | -202.8kb | Sense | Chr. 18 - 79144752 - 79149853 on NCBI Build GRCh38 | |
| Downstream | nuclear factor of activated T-cells 1 | NFATC1 |
NF-ATC, NF-ATc1.2, NFAT2, NFATc |
41.2kb | Sense | Chr. 18 - 79395772 - 79529323 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK130839.1 transcripts.
Set Membership: |
|

ATP9B
Genomic Map