Assay Details
| Gene Symbol: LOC100128170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124170.1 | - | 1 | 1843 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105372626 | LOC105372626 | -59.1kb | Sense | Chr. 20 - 43894718 - 43895468 on NCBI Build GRCh38 | |
| Downstream | junctophilin 2 | JPH2 |
CMH17, JP-2, JP2 |
155.1kb | Anti-sense | Chr. 20 - 44111697 - 44187578 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100128170
Genomic Map