Assay Details
| Gene Symbol: LOC100130463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124300.1 | - | 1 | 1154 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4502 | MIR4502 | -18.4kb | Sense | Chr. 13 - 114273828 - 114273908 on NCBI Build GRCh38 | |
| Downstream | chromosome alignment maintaining phosphoprotein 1 | CHAMP1 |
C13orf8, CAMP, CHAMP, MRD40, ZNF828 |
19.8kb | Sense | Chr. 13 - 114314490 - 114327328 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100130463
Genomic Map