Assay Details
| Gene Symbol: LOC727808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124942.1 | - | 1 | 650 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100130111 | LOC100130111 | -141.4kb | Sense | Chr. 15 - 29674935 - 29679163 on NCBI Build GRCh38 | |
| Downstream | golgin A8 family member J | GOLGA8J | 258.3kb | Sense | Chr. 15 - 30082627 - 30096711 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC727808
Genomic Map