| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008574.hg.1 |
TC08000622.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008574.hg.1 |
ENST00000514926.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008574.hg.1 |
NR_033962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008573.hg.1 |
TC08000622.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008573.hg.1 |
ENST00000514926.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008573.hg.1 |
NR_033962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008396.hg.1 |
ENST00000514926.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008396.hg.1 |
ENST00000514926.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008396.hg.1 |
ENST00000514926.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008396.hg.1 |
NR_033962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008396.hg.1 |
NR_033962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008396.hg.1 |
NR_033962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157783.hg.1 |
ENST00000514926.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157783.hg.1 |
NR_033962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157783.hg.1 |
ENST00000514926.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157783.hg.1 |
NR_033962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157783.hg.1 |
TC0800008396.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800008396.hg.1 |
ENST00000514926.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008396.hg.1 |
ENST00000514926.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008396.hg.1 |
ENST00000514926.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008396.hg.1 |
NR_033962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008396.hg.1 |
NR_033962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008396.hg.1 |
NR_033962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157784.hg.1 |
ENST00000514926.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157784.hg.1 |
NR_033962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157784.hg.1 |
ENST00000514926.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157784.hg.1 |
NR_033962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157784.hg.1 |
TC0800008396.hg.1 |
Clariom D TC |