Assay Details
| Gene Symbol: LOC100129484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125308.1 | - | 1 | 2453 | 93 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BM668076.1 | - | 1 | Chr. 7 - 5426277 - 5426606 [+] on Build GRCh38 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | trinucleotide repeat containing 18 | TNRC18 |
CAGL79, TNRC18A |
-2.7kb | Sense | Chr. 7 - 5306790 - 5426043 on NCBI Build GRCh38 |
| Downstream | F-box and leucine rich repeat protein 18 | FBXL18 | Fbl18 | 25.5kb | Sense | Chr. 7 - 5454448 - 5513813 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100129484
Genomic Map