Assay Details
| Gene Symbol: LOC100131820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125613.1 | - | 1 | 1744 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SWAP switching B-cell complex 70kDa subunit | SWAP70 |
HSPC321, SWAP-70 |
Sense | Chr. 11 - 9664077 - 9752991 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC440028 | LOC440028 | 37.1kb | Anti-sense | Chr. 11 - 9754770 - 9759533 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100131820
Genomic Map