Assay Details
| Gene Symbol: FLNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125630.1 | - | 1 | 1757 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105373386 | LOC105373386 | -30.0kb | Sense | Chr. X - 154333699 - 154347906 on NCBI Build GRCh38 | |
| Downstream | emerin | EMD |
EDMD, LEMD5, STA |
4.6kb | Anti-sense | Chr. X - 154379237 - 154381523 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

FLNA
Genomic Map