Assay Details
| Gene Symbol: LOC642422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125752.1 | - | 2 - 3 | 1437 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 200 | LINC00200 |
C10orf139, NCRNA00200 |
-173.1kb | Anti-sense | Chr. 10 - 1159768 - 1164672 on NCBI Build GRCh38 |
| Downstream | ADARB2 antisense RNA 1 | ADARB2-AS1 |
C10orf109, NCRNA00168, bA466B20.1 |
186.4kb | Anti-sense | Chr. 10 - 1526630 - 1556984 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC642422
Genomic Map