Assay Details
| Gene Symbol: LOC100130778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125937.1 | - | 1 | 1446 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | adhesion G protein-coupled receptor D1 | ADGRD1 |
GPR133, PGR25 |
Anti-sense | Chr. 12 - 130953907 - 131141469 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1257 | LINC01257 | 140.3kb | Anti-sense | Chr. 12 - 131165011 - 131212931 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100130778
Genomic Map