Assay Details
| Gene Symbol: LOC100131048 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125976.1 | - | 1 - 2 | 2226 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC151121 | LOC151121 | -529.6kb | Sense | Chr. 2 - 129242173 - 129273891 on NCBI Build GRCh38 | |
| Downstream | placenta specific 9 pseudogene | LOC389033 | 116.1kb | Sense | Chr. 2 - 129922862 - 129934317 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477704 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593446 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477710 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593463 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593471 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477715 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593439 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477698 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477749 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593476 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477702 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593458 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477743 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477699 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15321791 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477712 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593464 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477765 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593493 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477711 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477748 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477689 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477716 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477757 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15321792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477760 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477770 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477721 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477745 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593455 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593467 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477688 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477718 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477753 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593411 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593442 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593441 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593452 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477713 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593443 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477762 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477766 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477740 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593453 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593402 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477764 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477709 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593437 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15321790 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477727 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477706 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593485 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593465 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477756 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477773 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593488 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593451 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477771 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477705 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593432 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477776 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593480 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593409 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477737 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477695 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477751 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477767 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477746 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477700 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477754 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15321793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477750 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477722 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477735 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21593444 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477761 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477691 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477690 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01477707 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3289441 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3289446 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 6748 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 52217 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3289446 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3289441 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P30712221 | - | - |

LOC100131048
Genomic Map