Assay Details
| Gene Symbol: LOC100131034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126091.1 | - | 1 | 2857 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kinesin family member 26A | KIF26A | -174.9kb | Anti-sense | Chr. 14 - 104138723 - 104180898 on NCBI Build GRCh38 | |
| Downstream | chromosome 14 open reading frame 180 | C14orf180 |
C14orf77, NRAC |
220.8kb | Anti-sense | Chr. 14 - 104579684 - 104589847 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09531010 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530932 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530950 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09531000 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16738934 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530939 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530940 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530970 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16738939 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530928 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994956 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530942 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29995009 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530941 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09531004 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530998 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994984 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29995015 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530974 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530931 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530978 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530908 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530972 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16738920 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530899 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530949 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09531007 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994964 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29995032 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530988 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530905 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530926 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530938 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09531008 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530925 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994983 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530933 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530921 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530947 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530995 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530903 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530922 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530909 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530917 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530923 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530904 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530985 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530956 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530953 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994961 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994966 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530969 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530981 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29995006 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530961 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530937 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29995036 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530994 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530901 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29995061 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530919 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530944 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530965 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530907 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29995072 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530906 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09531006 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530984 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09531009 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530920 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530929 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994990 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530999 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29995055 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994972 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09531001 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994945 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530989 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530912 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530943 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16738938 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530964 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16738935 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994975 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530934 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530977 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530959 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530946 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16738931 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530914 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29995051 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530952 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 812289 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 240796 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3296841 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 29000 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3296841 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02989582 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1910322 | AK126091.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1910322 | AK126091.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1910322 | AK126091.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_104784 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_104784 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1910322 | AK126091.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1910322 | AK126091.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1910322 | AK126091.1 | human_transcriptome |

LOC100131034
Genomic Map