Assay Details
Gene Symbol: LOC100129042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK126562.1 | - | 1 | 1650 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
kazrin, periplakin interacting protein |
KAZN | KAZ | Sense | Chr. 1 - 13893387 - 15118048 on NCBI Build GRCh38 | |
Downstream | TMEM51 antisense RNA 1 | TMEM51-AS1 | C1orf126 | 110.5kb | Anti-sense | Chr. 1 - 15111815 - 15152464 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
