Assay Details
| Gene Symbol: LOC100131395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127105.1 | - | 1 | 452 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 596 | MIR596 |
MIRN596, hsa-mir-596 |
-117.4kb | Anti-sense | Chr. 8 - 1817231 - 1817307 on NCBI Build GRCh38 |
| Downstream | kelch repeat and BTB domain containing 11 | KBTBD11 | KLHDC7C | 32.5kb | Anti-sense | Chr. 8 - 1970718 - 2006943 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100131395
Genomic Map