Assay Details
| Gene Symbol: LOC100131129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127184.1 | - | 1 | 2052 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cytochrome c oxidase subunit 4I1 | COX4I1 |
COX IV-1, COX4, COX4-1, COXIV, COXIV-1 |
-52.5kb | Sense | Chr. 16 - 85799567 - 85807003 on NCBI Build GRCh38 |
| Downstream | interferon regulatory factor 8 | IRF8 |
H-ICSBP, ICSBP, ICSBP1, IMD32A, IMD32B, IRF-8 |
37.2kb | Sense | Chr. 16 - 85899168 - 85922606 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709768 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167107 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709759 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167133 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709822 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167167 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709786 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922332 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167159 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709831 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709820 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167140 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167114 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167123 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167141 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709757 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167165 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709821 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167151 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922333 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709754 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167115 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167190 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709828 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167186 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167152 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167119 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709763 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167116 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167121 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167154 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709737 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167137 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167111 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167144 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167122 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167155 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167147 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167105 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167124 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167164 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922341 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709787 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709748 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167130 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709825 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922331 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167170 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709816 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922345 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167110 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167162 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167143 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167187 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167125 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709829 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167160 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709796 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167193 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709769 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709745 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709746 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167168 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167149 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922338 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709817 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709832 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167108 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167132 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709740 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709813 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709764 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167189 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167178 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30709809 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922351 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167146 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10167176 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16922340 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 47185 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3393456 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 26900 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3393456 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03193621 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008711.hg.1 | ENST00000624587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008711.hg.1 | ENST00000624587.1 | Ensembl |

LOC100131129
Genomic Map