Assay Details
| Gene Symbol: LOC401557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046107.1 | 2 - 3 | 701 | 59 | ||
| GenBank mRNA | AK127261.1 | - | 2 - 3 | 701 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | olfactomedin 1 | OLFM1 |
AMY, NOE1, NOELIN1, OlfA |
-62.7kb | Sense | Chr. 9 - 135075243 - 135121184 on NCBI Build GRCh38 |
| Downstream | chromosome 9 open reading frame 62 | C9orf62 | 155.9kb | Sense | Chr. 9 - 135343249 - 135346562 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000791.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352685 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002957 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003098 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352659 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352574 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352624 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352630 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002992 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002974 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352668 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352673 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003017 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352645 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352649 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352644 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003039 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003057 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002994 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352704 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002993 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002997 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002953 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003002 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003009 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002954 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003003 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003015 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352688 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003078 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352610 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352721 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352580 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002959 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003082 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352689 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002985 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352642 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003006 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002968 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002944 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002995 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002967 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352716 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003045 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003030 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003055 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003076 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003092 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002973 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003025 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003095 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003093 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003033 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352699 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003028 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352702 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003026 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002998 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003074 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352647 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003054 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003013 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352690 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002965 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003071 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002979 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002982 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003073 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003094 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003011 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002983 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352738 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003101 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002951 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002956 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003048 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003060 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002961 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003000 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003103 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002964 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003079 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352572 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002981 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002987 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003044 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003086 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002990 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003062 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003087 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003037 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003016 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003072 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352621 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002991 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002966 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352740 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002958 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003035 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003014 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003032 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002980 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003001 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002977 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002969 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003068 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002984 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003083 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003020 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003104 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003059 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352619 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003091 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352591 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352679 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352602 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003018 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003058 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352571 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002970 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003099 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003023 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352731 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352582 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002955 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352677 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003070 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003066 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003102 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002976 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003034 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003043 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352744 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003063 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003081 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352678 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003085 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002962 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07002949 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27352687 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07003046 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3365392 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3365387 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 62072 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 23643 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P02192415 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02192415 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 48755 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 34176 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32574262 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02192404 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32574329 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02192415 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32574398 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | XM_001719680_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8159236 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8159237 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8159238 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8159235 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000513384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000513384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000513384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000513384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | ENST00000513384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | NR_125911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | NR_125911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | NR_125911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | NR_125911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | NR_125911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010103.hg.1 | NR_125911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09010296.hg.1 | TC09000791.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09010296.hg.1 | NR_046107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09010295.hg.1 | TC09000791.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09010295.hg.1 | NR_046107.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09010297.hg.1 | TC09000791.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09010297.hg.1 | NR_046107.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000502344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000513384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000513384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000513384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000513384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | ENST00000513384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | NR_125911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | NR_125911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | NR_125911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | NR_125911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | NR_125911.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010103.hg.1 | NR_125911.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900162318.hg.1 | NR_046107.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900162318.hg.1 | NR_046107.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900162318.hg.1 | TC0900009148.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900162317.hg.1 | NR_046107.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900162317.hg.1 | NR_046107.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900162317.hg.1 | TC0900009148.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900162316.hg.1 | NR_046107.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900162316.hg.1 | NR_046107.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900162316.hg.1 | TC0900009148.hg.1 | Clariom D TC |

LOC401557
Genomic Map