Assay Details
| Gene Symbol: C5orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127800.1 | - | 1 | 5052 | 90 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DB327332.1 | - | 1 | Chr. 5 - 61703354 - 61703894 [+] on Build GRCh38 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928651 | LOC101928651 | -4.9kb | Sense | Chr. 5 - 61663157 - 61698432 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100506526 | LOC100506526 | 24.2kb | Sense | Chr. 5 - 61732790 - 61735698 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C5orf64
Genomic Map