Assay Details
| Gene Symbol: LOC100130768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128224.1 | - | 1 | 3249 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heparan sulfate 6-O-sulfotransferase 1 | HS6ST1 |
HH15, HS6ST |
-56.9kb | Sense | Chr. 2 - 128265480 - 128318597 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927881 | LOC101927881 | 485.6kb | Anti-sense | Chr. 2 - 128864600 - 128868727 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472546 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472536 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472639 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15320993 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588257 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472513 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472587 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588221 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15320979 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472601 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472512 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472530 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472526 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472652 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472578 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472648 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472622 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472605 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472617 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588280 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472568 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472629 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472604 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472514 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588235 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472518 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472541 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472632 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472607 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588281 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15320980 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472557 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472509 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588159 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472589 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472506 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472540 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472539 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472599 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472567 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472552 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472594 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472651 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15321009 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472580 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472569 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472585 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472563 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472624 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588232 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472631 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472636 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472529 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472586 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472646 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472551 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472566 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472582 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472576 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472571 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472538 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472570 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472634 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472610 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15320978 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472613 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472560 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472649 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472626 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472508 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472525 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472641 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472507 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472554 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472584 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472608 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472524 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472572 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472533 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588250 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472600 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15320984 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472633 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588256 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472630 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588207 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472650 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588233 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472602 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472520 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472511 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472606 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472556 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588279 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588216 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472555 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472505 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472516 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472615 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472640 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472545 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472517 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472611 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472527 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472618 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472548 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472581 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472535 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472543 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01472603 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588191 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588201 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21588153 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 107634 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 340205 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 253577 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3385681 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4554 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3385681 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 97569 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00466481 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00466481 | - | - |

LOC100130768
Genomic Map