Assay Details
| Gene Symbol: FLJ46552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128409.1 | - | 1 | 3350 | 66 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | HY200193.1 | - | 1 | Chr. 6 - 1617289 - 1618801 [+] on Build GRCh38 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | forkhead box C1 | FOXC1 |
ARA, FKHL7, FREAC-3, FREAC3, IGDA, IHG1, IRID1, RIEG3 |
-3.4kb | Anti-sense | Chr. 6 - 1610446 - 1613897 on NCBI Build GRCh38 |
| Downstream | GDP-mannose 4,6-dehydratase | GMDS |
GMD, SDR3E1 |
3.0kb | Sense | Chr. 6 - 1623800 - 2245634 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

FLJ46552
Genomic Map