Assay Details
| Gene Symbol: LOC100130931 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128535.1 | - | 1 | 952 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cyclin dependent kinase 5 regulatory subunit 1 | CDK5R1 |
CDK5P35, CDK5R, NCK5A, p23, p25, p35, p35nck5a |
-84.0kb | Sense | Chr. 17 - 32485281 - 32491256 on NCBI Build GRCh38 |
| Downstream | transmembrane protein 98 | TMEM98 | TADA1 | 350.7kb | Sense | Chr. 17 - 32927910 - 32941649 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100130931
Genomic Map