Assay Details
| Gene Symbol: LOC100128703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128830.1 | - | 44 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | collagen alpha-2(IV) chain-like | LOC101059906 | Sense | Chr. 11 - 528484 - 532263 on NCBI Build GRCh38 | ||
| Downstream |
HRas proto-oncogene, GTPase |
HRAS |
C-BAS/HAS, C-H-RAS, C-HA-RAS1, CTLO, H-RASIDX, HAMSV, HRAS1, RASH1, p21ras |
2.6kb | Anti-sense | Chr. 11 - 532242 - 535567 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK128830.1 transcripts.
Set Membership: |
|

LOC100128703
Genomic Map