Assay Details
| Gene Symbol: LOC100132368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131565.1 | - | 1 | 2554 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 614 | MIR614 |
MIRN614, hsa-mir-614 |
-57.5kb | Sense | Chr. 12 - 12915829 - 12915918 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100506314 | LOC100506314 | 3.8kb | Sense | Chr. 12 - 12979837 - 12984642 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100132368
Genomic Map