Assay Details
| Gene Symbol: LPEQ6126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY358194.1 | - | 1 | 942 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TBC1D22A antisense RNA 1 | TBC1D22A-AS1 | -199.8kb | Sense | Chr. 22 - 46914092 - 46916042 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC339685 | LOC339685 | 207.4kb | Anti-sense | Chr. 22 - 47324623 - 47366807 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LPEQ6126
Genomic Map