Assay Details
| Gene Symbol: TP53TG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC068535.1 | - | 1 | 2151 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 243 | TMEM243 |
C7orf23, MM-TRAG, MMTRAG |
-121.9kb | Sense | Chr. 7 - 87195593 - 87220603 on NCBI Build GRCh38 |
| Downstream | carnitine O-octanoyltransferase | CROT | COT | 109.0 | Anti-sense | Chr. 7 - 87345635 - 87399796 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TP53TG1
Genomic Map