Assay Details
| Gene Symbol: LOC645967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134652.1 | 1 | 1129 | 101 | ||
| GenBank mRNA | AB593152.1 | - | 1 | 1129 | 101 | |
| CA395838.1 | - | 1 | 385 | 101 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | large tumor suppressor kinase 1 | LATS1 |
WARTS, wts |
-161.0 | Sense | Chr. 6 - 149658153 - 149718256 on NCBI Build GRCh38 |
| Downstream | nucleoporin 43 | NUP43 |
bA350J20.1, p42 |
5.4kb | Sense | Chr. 6 - 149724315 - 149746572 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC645967
Genomic Map