Assay Details
| Gene Symbol: VPS35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF086051.1 | - | 1 | 453 | 70 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | N51700.1 | - | 1 | Chr. 16 - 46673443 - 46673927 [+] on Build GRCh38 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SHC binding and spindle associated 1 | SHCBP1 | PAL | -50.6kb | Sense | Chr. 16 - 46578591 - 46624933 on NCBI Build GRCh38 |
| Downstream | origin recognition complex subunit 6 | ORC6 | ORC6L | 15.7kb | Anti-sense | Chr. 16 - 46689646 - 46698394 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

VPS35
Genomic Map