Assay Details
| Gene Symbol: LINC00997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036501.1 | 1 | 1049 | 120 | ||
| GenBank mRNA | AF143329.1 | - | 1 | 152 | 120 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 550b-2 | MIR550B2 | -26.1kb | Anti-sense | Chr. 7 - 32732981 - 32733077 on NCBI Build GRCh38 | |
| Downstream | DPY19L1 pseudogene 2 | DPY19L1P2 | THC1072510 | 12.1kb | Sense | Chr. 7 - 32826302 - 32838744 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000210.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15924960 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25623404 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25623405 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05405083 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05405082 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25623438 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25623413 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P913716 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P457784 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 232255_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1560371_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 232256_PM_s_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011731.hg.1 | ENST00000471553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011731.hg.1 | ENST00000471553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007126.hg.1 | ENST00000609151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007126.hg.1 | ENST00000609151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007126.hg.1 | ENST00000628855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007126.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007126.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007126.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007126.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007126.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003310.hg.1 | TC07000210.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003310.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149602.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149602.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149602.hg.1 | TC0700007126.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700149604.hg.1 | ENST00000609151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149604.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149604.hg.1 | ENST00000609151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149604.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149604.hg.1 | TC0700007126.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700149606.hg.1 | ENST00000628855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149606.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149606.hg.1 | ENST00000628855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149606.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149606.hg.1 | TC0700007126.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700007126.hg.1 | ENST00000609151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007126.hg.1 | ENST00000609151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007126.hg.1 | ENST00000628855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007126.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007126.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007126.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007126.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007126.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149603.hg.1 | ENST00000609151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149603.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149603.hg.1 | ENST00000609151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149603.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149603.hg.1 | TC0700007126.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700011731.hg.1 | ENST00000471553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011731.hg.1 | ENST00000471553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700149605.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149605.hg.1 | NR_036501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700149605.hg.1 | TC0700007126.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1560371_at | - | - |
| Human Genome U133 Plus 2.0 Array | 232255_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 232256_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1560371_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 232255_at | - | - |
| Human Genome U133 Plus 2.0 Array | 232256_PM_s_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232256_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560371_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232255_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560371_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560371_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560371_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232256_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560371_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232256_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232256_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232255_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232255_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232256_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232256_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232256_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232255_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560371_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232256_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560371_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232255_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232255_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232255_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560371_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560371_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232255_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560371_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232255_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232255_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232256_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232255_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232256_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232256_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560371_at7 | - | - |
| Human Genome U95 Set | 47345_PM_r_at | - | - |
| Human Genome U95 Set | 47345_r_at | - | - |
| Human Genome U133 Set | 232255_PM_at | - | - |
| Human Genome U133 Set | 232256_PM_s_at | - | - |
| Human Genome U133 Set | 232256_s_at | - | - |
| Human Genome U133 Set | 232255_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034165 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034165 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1751227 | AK092714.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1751227 | AK127989.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1751227 | NR_036501.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1751227 | AK092714.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1751227 | AK127989.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1751227 | NR_036501.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1751227 | AK092714.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1751227 | AK127989.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1751227 | NR_036501.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1751227 | AK092714.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1751227 | AK127989.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1751227 | NR_036501.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1751227 | AK092714.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1751227 | AK127989.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1751227 | NR_036501.1 | human_transcriptome |

LINC00997
Genomic Map