Assay Details
| Gene Symbol: HFE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011514543.2 | XP_011512845.1 | 7 | 2735 | 86 | |
| GenBank mRNA | AF147392.1 | - | 1 | 441 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
histone cluster 1, H1c |
HIST1H1C |
H1.2, H1C, H1F2, H1s-1 |
-39.1kb | Anti-sense | Chr. 6 - 26055740 - 26056471 on NCBI Build GRCh38 |
| Downstream |
histone cluster 1, H4c |
HIST1H4C |
H4/g, H4FG, dJ221C16.1 |
7.9kb | Sense | Chr. 6 - 26103948 - 26104337 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

HFE
Genomic Map