Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PLCE1 antisense RNA 2 | PLCE1-AS2 | -171.1kb | Sense | Chr. 10 - 94081950 - 94108794 on NCBI Build GRCh38 | |
| Downstream | NOC3 like DNA replication regulator | NOC3L |
AD24, C10orf117, FAD24 |
31.4kb | Sense | Chr. 10 - 94333226 - 94362976 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map