Assay Details
| Gene Symbol: DEFB117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ012021.1 | - | 1 | 60 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | defensin beta 116 | DEFB116 | DEFB-16 | -52.2kb | Anti-sense | Chr. 20 - 31303212 - 31308585 on NCBI Build GRCh38 |
| Downstream | defensin beta 118 | DEFB118 |
C20orf63, DEFB-18, ESC42, ESP13.6 |
7.7kb | Sense | Chr. 20 - 31368618 - 31373923 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DEFB117
Genomic Map