Assay Details
| Gene Symbol: GHRHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | S79912.1 | - | 1 | 452 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | aquaporin 1 (Colton blood group) | AQP1 |
AQP-CHIP, CHIP28, CO |
-50.4kb | Sense | Chr. 7 - 30911800 - 30925516 on NCBI Build GRCh38 |
| Downstream | ADCYAP receptor type I | ADCYAP1R1 |
PAC1, PAC1R, PACAPR, PACAPRI |
75.9kb | Sense | Chr. 7 - 31052341 - 31114005 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GHRHR
Genomic Map