Assay Details
| Gene Symbol: MECOM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | S82592.1 | - | 1 | 356 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100507661 | LOC100507661 | -166.2kb | Anti-sense | Chr. 3 - 168901945 - 168921996 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105374205 | LOC105374205 | 358.7kb | Anti-sense | Chr. 3 - 169447867 - 169477052 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MECOM
Genomic Map