Assay Details
Gene Symbol: LOC100132661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF130044.2 | - | 1 | 1953 | 102 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chromosome 3 open reading frame 58 | C3orf58 |
DIA1, GoPro49, HASF |
-225.1kb | Anti-sense | Chr. 3 - 143971798 - 143992368 on NCBI Build GRCh38 |
Downstream | procollagen-lysine,2-oxoglutarate 5-dioxygenase 2 | PLOD2 |
BRKS2, LH2, TLH |
1.8mb | Sense | Chr. 3 - 146069436 - 146161495 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782587 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782617 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782553 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02641965 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782601 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782551 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02641992 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782548 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02642062 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02641973 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02641974 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782591 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02642045 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782583 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782622 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782570 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782546 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782573 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782589 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782605 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782602 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782582 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02642042 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02641964 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782618 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02642011 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02642020 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782585 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02642054 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782584 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782574 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782542 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02642013 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02641970 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782609 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02641966 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02642065 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782614 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782566 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782569 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782562 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02642018 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782564 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782621 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782596 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782586 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782590 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782608 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782549 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782616 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782597 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782558 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782552 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02642017 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02641976 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782554 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782576 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782555 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02642015 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782615 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782540 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02642047 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782598 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782594 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782563 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782599 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782568 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782604 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782557 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02642002 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782556 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22782536 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02641989 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 313766 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P145414 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 3954 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P145414 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P788373 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P145414 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 224103_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 224103_at | - | - |
Human Genome U133 Plus 2.0 Array | 224103_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 224103_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 224103_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 224103_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 224103_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 224103_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 224103_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 224103_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 224103_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 224103_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 224103_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 224103_at4 | - | - |
Human Genome U133 Set | 224103_PM_at | - | - |
Human Genome U133 Set | 224103_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina human-6 v2.0 expression beadchip | ILMN_1870953 | - | - |
Illumina Sentrix Human-6 v2 Expression BeadChip | 7100521 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_89001 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1886177 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1886177 | - | - |
Illumina HumanWG-6_V2_0_R2 | ILMN_1870953 | - | - |
