| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
XM_001725248_at |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8003059 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8003057 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8003058 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8003056 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1555645_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11761566_PM_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11761566_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010985.hg.1 |
ENST00000624764.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010985.hg.1 |
ENST00000624764.1 |
Ensembl |
| Human Genome U133 Plus 2.0 Array |
1555645_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1555645_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555645_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555645_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555645_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555645_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555645_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555645_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555645_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555645_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555645_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555645_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555645_at3 |
- |
- |