Assay Details
Gene Symbol: CECR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF307449.1 | - | 1 | 53 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
cat eye syndrome chromosome region, candidate 3 (non-protein coding) |
CECR3 | -62.3kb | Anti-sense | Chr. 22 - 17256859 - 17266733 on NCBI Build GRCh38 | |
Downstream |
CECR2, histone acetyl-lysine reader |
CECR2 | 30.7kb | Sense | Chr. 22 - 17359949 - 17558155 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091989 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091992 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11423700 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091986 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091968 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091994 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091947 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091991 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11423703 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091984 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091980 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11423719 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11423701 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11423715 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091952 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17251488 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091953 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11423717 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11423707 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091971 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17251487 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091951 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11423709 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11423713 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11423721 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11423690 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091990 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091960 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11423696 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091956 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091967 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32091950 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11423698 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3631491 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 53454 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3631491 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1567687_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1567686_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC2200006516.hg.1 | ENST00000453421.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006516.hg.1 | ENST00000453421.1 | Ensembl |
Human Genome U133 Plus 2.0 Array | 1567687_at | - | - |
Human Genome U133 Plus 2.0 Array | 1567686_at | - | - |
Human Genome U133 Plus 2.0 Array | 1567686_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1567687_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567687_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567686_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567686_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567686_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567686_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567686_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567686_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567687_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567687_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567686_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567687_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567687_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567687_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567686_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567687_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567687_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567686_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567687_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567686_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567687_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567686_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1567687_at9 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina human-6 v2.0 expression beadchip | ILMN_1844024 | - | - |
Illumina Sentrix Human-6 v2 Expression BeadChip | 5890068 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_106520 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1844024 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1844024 | - | - |
Illumina HumanWG-6_V2_0_R2 | ILMN_1844024 | - | - |
