Assay Details
| Gene Symbol: LOC93444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK000175.1 | - | 1 | 1949 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | prostate cancer associated transcript 29 (non-protein coding) | PCAT29 | -109.1kb | Sense | Chr. 15 - 69592200 - 69695750 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 593 | LINC00593 | C15orf50 | 28.3kb | Sense | Chr. 15 - 69835234 - 69842967 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232638 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739325 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16801591 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739273 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739374 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739375 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739346 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739277 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739388 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739379 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739345 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739265 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739274 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739267 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739366 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739278 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16801579 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232580 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739317 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739326 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16801585 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739387 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739334 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16801588 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739289 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739311 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16801577 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739287 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739340 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739291 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739323 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739343 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739361 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739285 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232578 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739389 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739364 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232656 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232531 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16801582 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232639 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739336 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739298 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739338 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739279 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739368 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16801597 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232608 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739363 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739378 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739299 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739356 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739286 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739396 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739393 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16801578 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739373 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739355 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232536 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232618 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739301 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232650 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232579 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739395 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739309 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232563 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739354 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232542 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739321 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739304 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739294 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739316 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739319 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739270 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232613 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739297 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739307 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739288 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232621 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16801599 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232555 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232523 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739296 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739327 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232649 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739394 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739358 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16801590 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739290 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739408 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739330 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739320 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739315 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232592 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739306 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739357 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739332 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232665 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739337 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739333 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232554 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739377 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739385 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739303 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739405 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739390 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739360 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739281 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739283 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232620 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232546 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739382 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232669 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739406 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739401 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739324 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739269 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739399 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739362 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739342 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232610 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739347 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739308 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232585 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232530 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739384 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30232643 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739275 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16801573 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739266 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16801589 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16801580 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09739380 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 781174 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3884005 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 44155 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3884005 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03055720 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 234956_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1564168_PM_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1564168_PM_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 234956_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 234956_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1564168_a_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234956_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234956_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564168_a_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564168_a_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564168_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234956_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564168_a_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234956_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564168_a_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564168_a_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234956_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564168_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564168_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234956_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564168_a_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564168_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234956_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234956_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564168_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234956_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234956_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234956_at3 | - | - |
| Human Genome U95 Set | 68379_at | - | - |
| Human Genome U95 Set | 68379_PM_at | - | - |
| Human Genome U133 Set | 234956_at | - | - |
| Human Genome U133 Set | 234956_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina human-6 v2.0 expression beadchip | ILMN_1863966 | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 7210193 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_84004 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1842251 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1842251 | - | - |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1863966 | - | - |

LOC93444
Genomic Map